Learn More
CD202b (TIE2) Monoclonal Antibody (TEK4), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Supplier: Invitrogen 14598781
Description
Description: The TEK4 monoclonal antibody reacts with mouse Tie-2, also known as CD202. A member of the tyrosine kinase receptor family, Tie-2 is expressed on endothelial and a subset of hematopoietic cells and is believed to play a role in both angiogenesis and hematopoiesis during development of the mouse embryo. In fetal liver and adult bone marrow, Tie-2 is expressed by a subpopulation of hematopoietic progenitor cells characterized as Lineage markers^-, c-Kit^+, Sca1^+ cells. Long-term multilineage repopulating cells were detected in Tie-2^+, Lineage^-, c-Kit^+, Sca1^+ cells but not in Tie-2^-, Lineage^-, c-Kit^+, Sca1^+ cells. Applications Reported: This TEK4 antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections. Applications Tested: The TEK4 antibody has been tested by blocking of staining with fluorochrome conjugated TEK4 on mouse bone marrow cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Filtration: 0.2 μm post-manufacturing filtered. The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells in mice, rats, and humans. This receptor possesses a unique extracellular domain containing 2 immunoglobulin-like loops separated by 3 epidermal growth factor-like repeats that are connected to 3 fibronectin type III-like repeats. The ligand for the receptor is angiopoietin-1. Defects in TEK are associated with inherited venous malformations; the TEK signaling pathway appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis. TEK is closely related to the TIE receptor tyrosine kinase.Specifications
CD202b (TIE2) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q02858 | |
Tek | |
Affinity chromatography | |
RUO | |
21687 | |
4° C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen) | |
TEK4 | |
Unconjugated | |
Tek | |
CD202B, TIE-2, TIE2, VMCM, VMCM1, angiopoietin-1 receptor, endothelial tyrosine kinase, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, tunica interna endothelial cell kinase, tyrosine kinase with Ig and EGF homology domains-2, tyrosine-protein kinase receptor TEK, tyrosine-protei | |
Rat | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
For Research Use Only.