missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD202b (TIE2) Rat anti-Mouse, Biotin, Clone: TEK4, eBioscience™

Rat Monoclonal Antibody

$143.88 - $566.28

Specifications

Antigen CD202b (TIE2)
Clone TEK4
Host Species Rat
Gene Alias TIE-2, TEK, CD202
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012099 Life Technologies
LS13598781
50μg Each for $143.88
5012100 Life Technologies
LS13598782
100μg Each for $262.68
5012101 Life Technologies
LS13598785
500μg Each for $566.28
Specifications

Description

The TEK4 monoclonal antibody reacts with mouse Tie-2, also known as CD202. A member of the tyrosine kinase receptor family, Tie-2 is expressed on endothelial and a subset of hematopoietic cells and is believed to play a role in both angiogenesis and hematopoiesis during development of the mouse embryo. In fetal liver and adult bone marrow, Tie-2 is expressed by a subpopulation of hematopoietic progenitor cells characterized as Lineage markers-, c-Kit+, Sca1+ cells. Long-term multilineage repopulating cells were detected in Tie-2+, Lineage-, c-Kit+, Sca1+ cells but not in Tie-2-, Lineage-, c-Kit+, Sca1+ cells.
Description & Specifications

Specifications

CD202b (TIE2)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TEK4
TIE-2, TEK, CD202
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL