missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD201 (EPCR) Rat anti-Mouse, Biotin, Clone: eBio1560 (1560), eBioscience™

Rat Monoclonal Antibody

$158.05 - $404.55

Specifications

Antigen CD201 (EPCR)
Clone eBio1560 (1560)
Host Species Rat
Gene Alias Endothelial Protein C Receptor
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011814 Life Technologies
LS13201280
25μg Each for $158.05
5011815 Life Technologies
LS13201282
100μg Each for $404.55
Specifications

Description

The eBio1560 monoclonal antibody reacts with mouse Endothelial Protein C Receptor (EPCR, CD201), a 25kDa Type 1 transmembrane protein expressed on endothelial cells. EPCR exhibits sequence and structural homology with the MHC class I/CD1 family of proteins. EPCR is a ligand for Protein C and plays an important role in augmenting Protein C activation by the thrombin-thrombomodulin complex and in regulating blood coagulation and inflammation. Deletion of EPCR results in embryonic lethality, at least partically due to placental thrombosis.

Recently, it was demonstrated that EPCR expression identified cells in the bone marrow that are capable of hematopoietic reconstitution activity comparable to hematopoietic stem cells isolated with conventional methods. The eBio1560 monoclonal antibody can be used for the detection of these hematopoietic stem cells, however the eBiomRCR-16 monoclonal antibody should only be used for the detection of CD201 on endothelial cells.
Description & Specifications

Specifications

CD201 (EPCR)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBio1560 (1560)
Endothelial Protein C Receptor
Flow Cytometry,Immunohistochemistry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL