missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD201 (EPCR) Rat anti-Human, APC, Clone: RCR-227, eBioscience™

Rat Monoclonal Antibody

$143.55 - $390.05

Specifications

Antigen CD201 (EPCR)
Clone RCR-227
Host Species Rat
Gene Alias Endothelial protein C receptor
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123099 Life Technologies
LS17201841
25 Tests Each for $143.55
501123100 Life Technologies
LS17201842
100 Tests Each for $390.05
Specifications

Description

The monoclonal antibody RCR-227 reacts with human CD201, which is also known as the endothelial protein C receptor (EPCR), a 46kDa transmembrane protein related to class I MHC. CD201 was originally identified on vascular endothelium cells, predominantly larger blood vessels. It is also expressed on monocytes, lymphocytes, hematopoietic stem cells, neutrophils, and eosinophils. CD201 serves as a binding site for protein C, activated protein C (APC), and factor VII/VIIa. A soluble form has also been identified in the plasma. CD201 plays a role in controlling coagulation as disruption of its gene in mice caused placental thrombosis and early embryonic death. This cell surface molecule is also involved in the anti-inflammatory response. APC binding to CD201 enables PAR-1 activation that confers cytoprotective effects. In monocytes, APC binding decreases secretion of proinflammatory cytokines, chemokines, and growth factors. Finally, CD201 has been shown to inhibit lymphocyte migration.
Description & Specifications

Specifications

CD201 (EPCR)
Rat
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
RCR-227
Endothelial protein C receptor
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test