Learn More
CD200 Receptor Monoclonal Antibody (OX108), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$542.00 - $542.00
Specifications
Antigen | CD200 Receptor |
---|---|
Clone | OX108 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
501128656
|
Invitrogen
46920142 |
100 Tests |
Each for $542.00
|
|
|||||
Description
Description: The monoclonal antibody OX108 recognizes human CD200R also known as OX2. CD200R is an inhibitory receptor with a similar structure as its ligand, CD200, yet lacks an ITIM domain typically found in inhibitory receptors. Instead the CD200R cytoplasmic domain contains a novel phosphotyrosine binding domain, NPXY, which after binding SHIP inhibits ERK, JNK and MAPK p38 pathways. In activated macrophages signaling results in inhibition of TNFalpha secretion. Isoforms of CD200R have been identified and are thought to play a major role in differentiation, especially in regards to tolerogenic DCs. Expression is restricted to hematopoietic cells: myeloid cells (monocytes, macrophages, DCs, neutrophils, mast cells and basophils) and a subset of T lymphocytes as well as langerhans (LC) cells and dendritic epidermal T cells. The epitope of OX108 is thought to be near the binding site of CD200. Applications Reported: This OX108 antibody has been reported for use in flow cytometric analysis. Applications Tested: This OX108 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD200 Receptor Monoclonal antibody specifically detects CD200 Receptor in Human samples. It is validated for Flow Cytometry.Specifications
CD200 Receptor | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q8TD46 | |
CD200R1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD200R1 |
OX108 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
131450 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.