Please login to your online account to display your discounted pricing

CD200 Rat anti-Mouse, PE, Clone: OX90, eBioscience™

Rat Monoclonal Antibody

$129.05 - $288.55

Specifications

Antigen CD200
Clone OX90
Host Species Rat
Gene Alias OX2, OX-2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010811

 
life technologies
LS12520080
25μg Each for $129.05

5010812

 
life technologies
LS12520082
100μg Each for $288.55
Description & Specifications

Specifications

Antigen CD200
Clone OX90
Host Species Rat
Gene Alias OX2, OX-2
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody OX90.1 recognizes mouse CD200 also known as OX2. CD200 is a member of the Ig superfamily with 2 Ig domains, a transmembrane and cytoplasmic domain. CD200 is expressed on resting and activated B cells, a subset of resting and activated T cells, keratinocytes, peripheral and central nerve cells, follicular dendritic cells and ovarian cells. The interaction with CD200R results in macrophage activation (IL-6 production), inhibition of mast cell degranulation along with reduced TN Falpha and IL-13 secretion and overall attenuation of the activation status of lymphocytes.