Please login to your online account to display your discounted pricing

CD200 Mouse anti-Human, PE, Clone: OX104, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $346.55

Specifications

Antigen CD200
Clone OX104
Host Species Mouse
Gene Alias OX-2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122429

 
life technologies
LS12920041
25 tests Each for $143.55

501130695

 
life technologies
LS12920042
100 tests Each for $346.55
Description & Specifications

Specifications

Antigen CD200
Clone OX104
Host Species Mouse
Gene Alias OX-2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The monoclonal antibody OX104 recognizes human CD200 also known as OX2. CD200 is a member of the Ig superfamily with 2 Ig domains, a transmembrane and cytoplasmic domain. CD200 is expressed on resting and activated B cells, a subset of resting and activated T cells, keratinocytes, peripheral and central nerve cells, follicular dendritic cells and ovarian cells. The interaction with CD200R results in macrophage activation (IL-6 production), inhibition of mast cell degranulation along with reduced TNF alpha and IL-13 secretion and overall attenuation of the activation status of lymphocytes. A role has also been suggested in maternal tolerance as expression of CD200 is also present on the trophoblast.