missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD20 Rat anti-Mouse, PE, Clone: QCH6A7, eBioscience™

Rat Monoclonal Antibody

$209.88 - $381.48

Specifications

Antigen CD20
Clone QCH6A7
Host Species Rat
Gene Alias Ms4a1, Ly-44
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122215 Life Technologies
LS12020380
25μg Each for $209.88
501122216 Life Technologies
LS12020382
100μg Each for $381.48
Specifications

Description

The monoclonal antibody QCH6A7 reacts with mouse CD20. CD20 is a B cell specific, 32kDa membrane protein that is expressed on late pro-B cells through memory B cells in the B cell lineage. CD20 is not found on hematopoietic stem cells, early pro-B cells or later mature plasma cells. CD20 is involved in intracellular calcium regulation and plays a role in B cell proliferation. CD20 is intially expressed after the induction of CD19 during development and is coexpressed with IgM during the pre-B to immature B cell transition in bone marrow. Expression of CD20 increases during maturation with almost all mature B cells expressing some level of CD20. Unlike other B cell antigens, CD20 is not shed or internalized upon antibody binding. Monoclonal antibodies against the CD20 antigen may be used to deplete B cells and CD20 has proven to be an effective immunotheraputic target in B cell lymphomas and several autoimmune diseases including rheumatoid arthritis and type I diabetes. CD20 consists of 4 transmembrane domains and a 44 amino acid extracellular domain which contains the QCH6A7 epitope.

Staining with clone QCH6A7 provides significantly improved staining intensity compared to clone AISB12 in flow cytometric assays.
Description & Specifications

Specifications

CD20
Rat
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
QCH6A7
Ms4a1, Ly-44
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer