missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD20 Mouse anti-Human, PE-eFluor® 610, Clone: 2H7, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS61020941

Catalog No. 501124743



Description

Description

Description: The 2H7 monoclonal antibody reacts with human CD20, a 33-36 kDa transmembrane protein. CD20 is expressed by developing B cells as well as mature B cells but not plasma cells. CD20 has been detected at low levels on a small subset of mature T

00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD20 is a non-glycosylated surface phosphoprotein that has a molecular weight range of 33-37 kDa depending on the degree of phosphorylation. CD20 is expressed on mature and most maligt B cells, in a subpopulation of T lymphocytes and follicular dendritic cells. CD20 expression on B cells is synchronous with the expression of surface IgM and it regulates transmembrane calcium conductance, cell cycle progression and B-cell proliferation. CD20 is also associated with lipid rafts, but the intensity of this association depends on extracellular triggering, employing CD20 conformational change, and/or BCR (B cell antigen receptor) aggregation. After the receptor ligation, BCR and CD20 colocalize and then rapidly dissociate before BCR endocytosis, whereas CD20 remains at the cell surface. CD20 serves as a useful target for antibody-mediated therapeutic depletion of B cells, as it is expressed at high levels on most B-cell maligcies, but does not become internalized or shed from the plasma membrane following monoclonal antibody treatment. Diseases associated with CD20 dysfunction include Ms4a1-related common variable immune deficiency.
Specifications

Specifications

CD20
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P11836
B1, Bp35, CD20, LEU-16, MGC3969, MS4A1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
2H7
PE-eFluor® 610
MS4A1
Liquid
MS4A1
IgG2b, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
931
Documents

For Research Use Only.