missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD20 Mouse anti-Human, PE, Clone: 2H7, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $275.88

Specifications

Antigen CD20
Clone 2H7
Host Species Murine
Gene Alias B1, Leu-16
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122217 Life Technologies
LS12020941
25 Tests Each for $117.48
50245261 Life Technologies
LS12020942
100 Tests Each for $275.88
Specifications

Description

The 2H7 monoclonal antibody reacts with human CD20, a 33-36kDa transmembrane protein. CD20 is expressed by developing B cells as well as mature B cells but not plasma cells. CD20 has been detected at low levels on a small subset of mature T cells. It is suggested that CD20 plays a role in B-cell activation.
Description & Specifications

Specifications

CD20
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2H7
B1, Leu-16
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test