Please login to your online account to display your discounted pricing

CD20 Mouse anti-Human, eVolve 655, Clone: 2H7, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $427.75

Specifications

Antigen CD20
Clone 2H7
Host Species Murine
Gene Alias B1, Leu-16
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125044

 
life technologies
LS86020941
25 Tests Each for $201.55

501125045

 
life technologies
LS86020942
100 Tests Each for $427.75
Description & Specifications

Specifications

Antigen CD20
Clone 2H7
Host Species Murine
Gene Alias B1, Leu-16
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eVolve 655
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL

The 2H7 monoclonal antibody reacts with human CD20, a 33-36kDa transmembrane protein. CD20 is expressed by developing B cells as well as mature B cells but not plasma cells. CD20 has been detected at low levels on a small subset of mature T cells. It is suggested that CD20 plays a role in B-cell activation.