Please login to your online account to display your discounted pricing

CD20 Mouse anti-Human, eFluor 450, Clone: 2H7, eBioscience™

Mouse Monoclonal Antibody

$176.90 - $397.30

Specifications

Antigen CD20
Clone 2H7
Host Species Mouse
Gene Alias B1, Leu-16
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016270

 
life technologies
LS48020941
25 tests Each for $176.90

5016271

 
life technologies
LS48020942
100 tests Each for $397.30
Description & Specifications

Specifications

Antigen CD20
Clone 2H7
Host Species Mouse
Gene Alias B1, Leu-16
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The 2H7 monoclonal antibody reacts with human CD20, a 33-36kDa transmembrane protein. CD20 is expressed by developing B cells as well as mature B cells but not plasma cells. CD20 has been detected at low levels on a small subset of mature T cells. It is suggested that CD20 plays a role in B-cell activation.