Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD20 Ab-1, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry, immunofluorescence and flow cytometry applications.

$270.94 - $975.33

Specifications

Antigen CD20 Ab-1
Clone L26
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS340R7

 
thermo scientific™ lab vision™
MS340R7
7mL Each for $471.06

MS340S

 
thermo scientific™ lab vision™
MS340S
1mL Each for $975.33

MS340S0

 
thermo scientific
MS340S0
100µL Each for $270.94

MS340S1

 
thermo scientific™ lab vision™
MS340S1
500µL Each for $648.18
Description & Specifications

Specifications

Antigen CD20 Ab-1
Clone L26
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Mouse
Regulatory Status IVD
Research Discipline B-Cell
Type Primary
Immunogen Human tonsil B cells

Obtain accurate and reproducible results in immunohistochemistry, immunofluorescence and flow cytometry applications with Thermo Scientific™ CD20 Ab-1, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

CD20 is a non-Ig differentiation antigen of B cells and the expression of CD20 is restricted to normal and neoplastic B cells, being absent from all other leukocytes and tissues. It acts as calcium channel involved in B cell activation and cell cycle progression.

Host Species: Mouse

Clone: L26

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with cow, dog, pig and rat.

Epitope: Cytoplasmic domain

Immunogen: Human tonsil B cells

Molecular Weight: 30-33kDa

Positive Control: Tonsil

Cellular Localization: Membrane/cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry