missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD2 Rat anti-Mouse, Biotin, Clone: RM2-5, eBioscience™

Rat Monoclonal Antibody

$104.28 - $341.88

Specifications

Antigen CD2
Clone RM2-5
Host Species Rat
Gene Alias LFA-2, Ly-37
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011365 Life Technologies
LS13002182
100μg Each for $196.68
5011364 Life Technologies
LS13002181
50μg Each for $104.28
5011366 Life Technologies
LS13002185
500μg Each for $341.88
Specifications

Description

The RM2-5 monoclonal antibody reacts with the mouse CD2 molecule, an approximately 50-55kDa cell surface receptor expressed by all mouse lymphocytes. Expression of the CD2 antigen in the mouse differs from that of the human in that B lineage cells in the mouse from the pre-B cell stage to mature B cells express CD2. CD2 is a ligand for CD48 in the mouse and is involved in adhesion and activation of T cells.
Description & Specifications

Specifications

CD2
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
RM2-5
LFA-2, Ly-37
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer