Please login to your online account to display your discounted pricing

CD2 Mouse anti-Human, Non-human primate, FITC, Clone: RPA-2.10, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $259.55

Specifications

Antigen CD2
Clone RPA-2.10
Host Species Mouse
Gene Alias LFA-2, Ly-37
Species Reactivity Baboon, Chimpanzee, Cynomolgus, Human, Pig, Rhesus Ma
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509352

 
life technologies
LS11002941
25 tests Each for $114.55

509353

 
life technologies
LS11002942
100 tests Each for $259.55
Description & Specifications

Specifications

Antigen CD2
Clone RPA-2.10
Host Species Mouse
Gene Alias LFA-2, Ly-37
Species Reactivity Baboon, Chimpanzee, Cynomolgus, Human, Pig, Rhesus Ma
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The RPA-2.10 monoclonal antibody reacts with human CD2, a 50kDa cell surface receptor expressed by a majority of thymocytes, all mature T cells and subset of NK cells. CD2 is a ligand for CD58 in the human and is involved in adhesion and activation of T cells. RPA-2.10 blocks mixed lymphocyte reaction.

RPA-2.10 crossreacts to non-human primates and pigs.