missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD2 Mouse anti-Human, Non-human primate, APC, Clone: RPA-2.10, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $355.08

Specifications

Antigen CD2
Clone RPA-2.10
Host Species Murine
Gene Alias LFA-2, Ly-37
Species Reactivity Baboon, Chimpanzee, Cynomolgus, Human, Pig, Rhesus Ma
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014834 Life Technologies
LS17002941
25 Tests Each for $157.08
5014835 Life Technologies
LS17002942
100 Tests Each for $355.08
Specifications

Description

The RPA-2.10 monoclonal antibody reacts with human CD2, a 50kDa cell surface receptor expressed by a majority of thymocytes, all mature T cells and subset of NK cells. CD2 is a ligand for CD58 in the human and is involved in adhesion and activation of T cells. RPA-2.10 blocks mixed lymphocyte reaction.

RPA-2.10 crossreacts to non-human primates and pigs.
Description & Specifications

Specifications

CD2
Murine
Baboon, Chimpanzee, Cynomolgus, Human, Pig, Rhesus Ma
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
RPA-2.10
LFA-2, Ly-37
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test