missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD1d Rat anti-Mouse, PE, Clone: 1B1, eBioscience™

Rat Monoclonal Antibody

$143.88 - $389.40

Specifications

Antigen CD1d
Clone 1B1
Host Species Rat
Gene Alias CD1.1, Ly-38
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010121 Life Technologies
LS12001181
50μg Each for $143.88
5010122 Life Technologies
LS12001182
100μg Each for $249.48
5010123 Life Technologies
LS12001183
200μg Each for $389.40
Specifications

Description

The 1B1 monoclonal antibody reacts with mouse CD1d, a 48kDa glycoprotein with structural homology to MHC class I molecules. While similar to MHC Class I, CD1d associates with beta2m, functionally CD1d is similar to MHC Class II. 1B1 detects CD1d at varying levels on mouse leukocytes. 1B1 detects beta2m associated CD1d.
Description & Specifications

Specifications

CD1d
Rat
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
1B1
CD1.1, Ly-38
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer