missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD1d Rat anti-Mouse, Clone: 1B1, eBioscience™

Rat Monoclonal Antibody

$85.55 - $361.05

Specifications

Antigen CD1d
Clone 1B1
Host Species Rat
Gene Alias CD1.1, Ly-38
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122628 Life Technologies
LS14001181
50μg Each for $85.55
5012324 Life Technologies
LS14001182
100μg Each for $129.05
5012325 Life Technologies
LS14001185
500μg Each for $361.05
Specifications

Description

The 1B1 monoclonal antibody reacts with mouse CD1d, a 48kDa glycoprotein with structural homology to MHC class I molecules. While similar to MHC Class I, CD1d associates with beta2m, functionally CD1d is similar to MHC Class II. 1B1 detects CD1d at varying levels on mouse leukocytes. 1B1 detects beta2m associated CD1d.
Description & Specifications

Specifications

CD1d
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
1B1
CD1.1, Ly-38
Flow Cytometry,Functional Assay,Immunohistochemistry,Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL