Please login to your online account to display your discounted pricing

CD1d Rat anti-Mouse, Alexa Fluor 488, Clone: 1B1, eBioscience™

Rat Monoclonal Antibody

$172.55 - $332.05

Specifications

Antigen CD1d
Clone 1B1
Host Species Rat
Gene Alias CD1.1, Ly-38
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124585

 
life technologies
LS53001182
100μg Each for $332.05

5016668

 
life technologies
LS53001180
25μg Each for $172.55
Description & Specifications

Specifications

Antigen CD1d
Clone 1B1
Host Species Rat
Gene Alias CD1.1, Ly-38
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 1B1 monoclonal antibody reacts with mouse CD1d, a 48kDa glycoprotein with structural homology to MHC class I molecules. While similar to MHC Class I, CD1d associates with beta2m, functionally CD1d is similar to MHC Class II. 1B1 detects CD1d at varying levels on mouse leukocytes. 1B1 detects beta2m associated CD1d.