Please login to your online account to display your discounted pricing

CD1c Mouse anti-Human, FITC, Clone: L161, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $317.55

Specifications

Antigen CD1c
Clone L161
Host Species Murine
Gene Alias BDCA-1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509344

 
life technologies
LS11001541
25 tests Each for $143.55

509345

 
life technologies
LS11001542
100 Tests Each for $317.55
Description & Specifications

Specifications

Antigen CD1c
Clone L161
Host Species Murine
Gene Alias BDCA-1
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunoprecipitation
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

This L161 monoclonal antibody detects CD1c (also known as BDCA-1), a glycoprotein that is noncovalently linked to beta-2 microglobulin on thymocytes and antigen presenting cells such as dendritic and Langerhans cells. This molecule is also expressed on some circulating and marginal zone B cells, as well as in lymph nodes and germinal centers. CD1c is involved in the presentation of lipid antigens such as microbial fatty acids to effector T cells during the adaptive immune response. Finally, alternative splicing gives rise to three different isoforms of CD1c (soluble, membrane, and cytoplasmic/soluble isoforms).