missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD1c Mouse anti-Human, APC, Clone: L161, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $332.05

Specifications

Antigen CD1c
Clone L161
Host Species Murine
Gene Alias BDCA-1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014830 Life Technologies
LS17001541
25 Tests Each for $172.55
5014831 Life Technologies
LS17001542
100 Tests Each for $332.05
Specifications

Description

This L161 monoclonal antibody detects CD1c (also known as BDCA-1), a glycoprotein that is noncovalently linked to beta-2 microglobulin on thymocytes and antigen presenting cells such as dendritic and Langerhans cells. This molecule is also expressed on some circulating and marginal zone B cells, as well as in lymph nodes and germinal centers. CD1c is involved in the presentation of lipid antigens such as microbial fatty acids to effector T cells during the adaptive immune response. Finally, alternative splicing gives rise to three different isoforms of CD1c (soluble, membrane, and cytoplasmic/soluble isoforms).
Description & Specifications

Specifications

CD1c
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
L161
BDCA-1
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test