Please login to your online account to display your discounted pricing

CD1b Mouse anti-Human, APC, Clone: eBioSN13, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $390.05

Specifications

Antigen CD1b
Clone eBioSN13 (SN13 K5-1B8)
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014832

 
affymetrix
17001841
25 tests Each for $172.55

5014833

 
affymetrix
17001842
100 tests Each for $390.05
Description & Specifications

Specifications

Antigen CD1b
Clone eBioSN13 (SN13 K5-1B8)
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The eBioSN13 monoclonal antibody reacts with human CD1b. The CD1 molecules are similar to MHC class I, but rather than presenting peptide antigens to T-cell receptors (TCR), they present a diversity of lipid and glycolipid antigens which are derived from self as well as from mycobacteria which include sphingolipids, mycolic acid derivatives and phosphatidylinositols such as liparabinomannan (LAM) and glucose monomycolate (GMM). CD1b is 45 kDa, and like the other CD1 antigens, is expressed as a heterodimer composed of the CD1b noncovalently paired with β2-microglobulin.