Please login to your online account to display your discounted pricing

CD1a Mouse anti-Human, PE, Clone: HI149, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $332.05

Specifications

Antigen CD1a
Clone HI149
Host Species Mouse
Gene Alias Leu-6, T6, R4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010128

 
affymetrix
12001941
25 tests Each for $143.55

5010129

 
affymetrix
12001942
100 tests Each for $332.05
Description & Specifications

Specifications

Antigen CD1a
Clone HI149
Host Species Mouse
Gene Alias Leu-6, T6, R4
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The HI149 monoclonal antibody reacts with human CD1a, a 49kDa protein expressed by cortical thymocytes and dendritic cells including Langerhans cells. The CD1 family of proteins share some structural and functional characteristics with the MHC class I molecules; however, members of the CD1 family are not polymorphic. Similar to MHC class I, CD1a associates with the beta2-microglobulin and is thought to play a role in antigen presentation.