missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD1a Mouse anti-Human, eFluor 450, Clone: HI149, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $355.08

Specifications

Antigen CD1a
Clone HI149
Host Species Murine
Gene Alias Leu-6, T6, R4
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124077 Life Technologies
LS48001942
100 Tests Each for $355.08
5016222 Life Technologies
LS48001941
25 Tests Each for $157.08
Specifications

Description

The HI149 monoclonal antibody reacts with human CD1a, a 49kDa protein expressed by cortical thymocytes and dendritic cells including Langerhans cells. The CD1 family of proteins share some structural and functional characteristics with the MHC class I molecules; however, members of the CD1 family are not polymorphic. Similar to MHC class I, CD1a associates with the beta2-microglobulin and is thought to play a role in antigen presentation.
Description & Specifications

Specifications

CD1a
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HI149
Leu-6, T6, R4
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test