Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD1a Ab-5, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD1a Ab-5, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1856P1ABX

 View more versions of this product

Catalog No. MS1856P1ABX

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2a kappa
Cross Reactivity Human
Clone O10
Host Species Mouse
Format Concentrated
Regulatory Status IVD
Quantity 100µL
Applications Immunohistochemistry (Paraffin)
Antigen CD1a Ab-5
Classification Monoclonal
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Concentration 1mg/mL
Type Primary
Research Discipline Hematopoietic

Obtain accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD1a Ab-5, Mouse Monoclonal Antibody.

At least five CD1 genes (CD1a, b, c, d, and e) have been identified. CD1 is expressed on cortical thymocytes, Langerhans cells, and dendritic cells. It is absent on mature peripheral blood T cells but intracytoplasmic expression is detected on activated T lymphocytes. CD1 proteins have been demonstrated to restrict T-cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation.

Host Species: Mouse

Clone: O10

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Molecular Weight: 49kDa

Positive Control: Tonsil

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)