Please login to your online account to display your discounted pricing

CD199 (CCR9) Mouse anti-Human, PE, Clone: BBC3M4, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $477.05

Specifications

Antigen CD199 (CCR9)
Clone BBC3M4
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122296

 
life technologies
LS12199941
25 Tests Each for $187.05

501122297

 
life technologies
LS12199942
100 Tests Each for $477.05
Description & Specifications

Specifications

Antigen CD199 (CCR9)
Clone BBC3M4
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This BBC3M4 monoclonal antibody reacts with human CD199 (CCR9), a 7 transmembrane spanning G-protein-coupled receptor. CCR9 expression is regulated during thymic development and CCR9 deficiency results in mild impairment in thymocyte development. In the periphery, CCR9 and alpha 4 beta 7 integrin are upregulated in response to dendritic cell-derived retinoic acid, conferring preferential trafficking to gut-associated tissues. CCR9 is the receptor for CCL25 (TECK).