missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD197 (CCR7), PE-eFluor 610, clone: 3D12, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS61197941

 View more versions of this product

Catalog No. 501124793



Description

Description

CCR7 is a member of the G protein coupled receptor family (subfamily: chemokine). This receptor was identified as a gene induced by the Epstein Barr virus (EBV), and is thought to be a mediator of EBV effects on B lymphocytes. CCR7 has been reported to be expressed in blood, bone marrow, lymph node, and intestine. It is particularly expressed in lymphoid tissues and in activated B and T lymphocytes and has been shown to control the migration of memory T cells to inflamed tissues, as well as stimulate dendritic cell maturation. The chemokine (C-C motif) ligand 19 (CCL19/ECL) has been reported to be a specific ligand of this receptor. ESTs have been isolated from blood, embryo, lymph node, and thymus libraries.

It is recommended that the staining incubation time be increased to at least 45 minutes at 2-8°C. PE-eFluor® 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red®). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CCR7 is a member of the G protein coupled receptor family, which is a subfamily of chemokines. CCR7 was identified to be induced by the Epstein Barr virus (EBV), and is thought to be a mediator of EBV effects on B lymphocytes. CCR7 has been reported to be expressed in blood, bone marrow, lymph node, and intestine. CCR7 is particularly expressed in lymphoid tissues and in activated B and T lymphocytes and has been shown to control the migration of memory T cells to inflamed tissues, as well as stimulate dendritic cell maturation. The chemokine (C-C motif) ligand 19 (CCL19/ECL) has been reported to be a specific ligand of this receptor. ESTs have been isolated from blood, embryo, lymph node, and thymus libraries. Receptors for the C - C chemokine family include CCR 1, CCR 2A, CCR 3, CCR 4, CCR 5 and the Duffy blood group antigen. The C-C receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation.
Specifications

Specifications

CD197 (CCR7)
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P32248
Bukitt′s lymphoma receptor 2; C-C chemokine receptor type 7; CC chemokine receptor 7; CCR-7; CD197; Chemokine (C-C Motif) Receptor 7; chemokine (C-C) receptor 7; CMKBR7; EBI1; EBV-induced G protein-coupled receptor 1; EBV-induced G-protein coupled receptor 1; Epstein-Barr virus induced gene 1; Epstein-Barr Virus-Induced G-Protein Coupled Receptor 1; lymphocyte-specific G protein-coupled peptide receptor; MIP-3 Beta Receptor
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
3D12
PE-eFluor® 610
CCR7
Liquid
CCR7
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
1236
Documents

For Research Use Only.