Please login to your online account to display your discounted pricing

CD197 (CCR7) Rat anti-Human, APC-eFluor 780, Clone: 3D12, eBioscience™

Rat Monoclonal Antibody

$216.05 - $506.05

Specifications

Antigen CD197 (CCR7)
Clone 3D12
Host Species Rat
Gene Alias EBI-1, MIP-3 beta Receptor
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016193

 
affymetrix
47197941
25 tests Each for $216.05

501129419

 
affymetrix
47197942
100 tests Each for $506.05
Description & Specifications

Specifications

Antigen CD197 (CCR7)
Clone 3D12
Host Species Rat
Gene Alias EBI-1, MIP-3 beta Receptor
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC-eFluor 780
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1μg)/test

The 3D12 monoclonal antibody reacts with human CCR7, also known as EBI-1 and CD197. CCR7 is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains and functions as a receptor for 6Ckine/SLC (secondary lymphoid-tissue chemokine), CCL19 and CCL21. CCR7 has been shown to be internalized via clathrin-coated pits and the majority recycled back to the plasma membrane. CCR7 is expressed on T cells and can be used to distinguish populations of naïve from central and effector memory T cells. CCR7 has been shown to play a role in migration of memory T cells to inflamed tissue. Expression of CCR7 is also found on DC’s. During DC maturation CCR7 expression increases and is thought to be involved in a variety of functions: chemotaxis to the lymph node, cellular architecture, rate of endocytosis, survival and maturation. Expression of CCR7 on the cell surface can be down regulated upon ligand binding.