Please login to your online account to display your discounted pricing

CD196 (CCR6) Mouse anti-Human, PerCP-eFluor 710, Clone: R6H1, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $506.05

Specifications

Antigen CD196 (CCR6)
Clone R6H1
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016021

 
life technologies
LS46196941
25 tests Each for $216.05

501130791

 
life technologies
LS46196942
100 tests Each for $506.05
Description & Specifications

Specifications

Antigen CD196 (CCR6)
Clone R6H1
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This R6H1 monoclonal antibody reacts with CD196 (also known as CCR6), a seven transmembrane G protein-coupled receptor expressed on T, B, dendritic, natural killer, and Langerhans cells. This CC chemokine receptor uniquely binds MIP-3a/CCL20, a chemoattractant for dendritic cells, effector/memory T cells, and B cells. CD196 is also involved in host defense and inflammation at epithelial sites. Furthermore, this receptor has been implicated in Th17 differentiation and CD4+FoxP3+ regulatory T cell development.