missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD196 (CCR6) Mouse anti-Human, Clone: R6H1, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $249.48

Specifications

Antigen CD196 (CCR6)
Clone R6H1
Host Species Murine
Applications Flow Cytometry,Immunoprecipitation,Western Blot
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012960 Life Technologies
LS14196980
25μg Each for $130.68
5012961 Life Technologies
LS14196982
100μg Each for $249.48
Specifications

Description

This R6H1 monoclonal antibody reacts with CD196 (also known as CCR6), a seven transmembrane G protein-coupled receptor expressed on T, B, dendritic, natural killer, and Langerhans cells. This CC chemokine receptor uniquely binds MIP-3a/CCL20, a chemoattractant for dendritic cells, effector/memory T cells, and B cells. CD196 is also involved in host defense and inflammation at epithelial sites. Furthermore, this receptor has been implicated in Th17 differentiation and CD4+FoxP3+ regulatory T cell development.
Description & Specifications

Specifications

CD196 (CCR6)
Murine
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
R6H1
Flow Cytometry,Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL