Please login to your online account to display your discounted pricing

CD195 (CCR5) Mouse anti-Human, PE, Clone: eBioT21/8 (T21/8), eBioscience™

Mouse Monoclonal Antibody

$158.05 - $419.05

Specifications

Antigen CD195 (CCR5)
Clone eBioT21/8 (T21/8)
Host Species Mouse
Gene Alias CCR-5
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010697

 
life technologies
LS12195741
25 tests Each for $158.05

5010698

 
life technologies
LS12195742
100 tests Each for $419.05
Description & Specifications

Specifications

Antigen CD195 (CCR5)
Clone eBioT21/8 (T21/8)
Host Species Mouse
Gene Alias CCR-5
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The eBioT21/8 monoclonal antibody reacts with the amino terminus of human CCR5 (CD195), an approximately 45kDa member of the G protein-coupled receptor (GPCR) superfamily that binds chemokines including MIP-1α, MIP-1β and RANTES. Human CCR5 is expressed on resting T cells with a memory/effector phenotype, monocytes, macrophages and immature dendritic cells. CCR5 functions as a receptor to direct the chemotaxis of lymphocytes, and is the primary co-receptor for macrophage-tropic Human Immunodeficiency Virus Type 1 (HIV-1), which binds CCR5 through gp120. Binding of the eBioT21/8 monoclonal antibody displays donor variability and may be affected by sub-optimal staining conditions.