Please login to your online account to display your discounted pricing

CD195 (CCR5) Mouse anti-Human, APC, Clone: NP-6G4, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $390.05

Specifications

Antigen CD195 (CCR5)
Clone NP-6G4
Host Species Murine
Gene Alias C-C chemokine receptor type 5, CMKBR5
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123098

 
life technologies
LS17195642
100 Tests Each for $390.05

501123097

 
life technologies
LS17195641
25 Tests Each for $187.05
Description & Specifications

Specifications

Antigen CD195 (CCR5)
Clone NP-6G4
Host Species Murine
Gene Alias C-C chemokine receptor type 5, CMKBR5
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The NP-6G4 monoclonal antibody reacts with human CD195 (CCR5), an approximately 45kDa member of the G protein-coupled receptor (GPCR) superfamily that binds chemokines including MIP-1a, MIP-1b and RANTES. Human CCR5 is expressed on resting T cells with a memory/effector phenotype, monocytes, macrophages and immature dendritic cells. CCR5 functions as a receptor to direct the chemotaxis of lymphocytes, and is the primary co-receptor for macrophage-tropic Human Immunodeficiency Virus Type 1 (HIV-1), which binds CCR5 through gp120.