Please login to your online account to display your discounted pricing

CD19 Rat anti-Mouse, PE-eFluor 610, Clone: eBio1D3 (1D3), eBioscience™

Rat Monoclonal Antibody

$158.05 - $390.05

Specifications

Antigen CD19
Clone eBio1D3 (1D3)
Host Species Rat
Gene Alias B4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124739

 
life technologies
LS61019380
25μg Each for $158.05

501124740

 
life technologies
LS61019382
100μg Each for $390.05
Description & Specifications

Specifications

Antigen CD19
Clone eBio1D3 (1D3)
Host Species Rat
Gene Alias B4
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBio1D3 (1D3) monoclonal antibody reacts with mouse CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells.