Please login to your online account to display your discounted pricing

CD19 Rat anti-Mouse, PE, Clone: eBio1D3 (1D3), eBioscience™

Rat Monoclonal Antibody

$143.55 - $796.05

Specifications

Antigen CD19
Clone eBio1D3 (1D3)
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010252

 
life technologies
LS12019381
50μg Each for $143.55

5010255

 
life technologies
LS12019385
500μg Each for $796.05

5010254

 
life technologies
LS12019383
200μg Each for $419.05

501129599

 
life technologies
LS12019382
100μg Each for $259.55
Description & Specifications

Specifications

Antigen CD19
Clone eBio1D3 (1D3)
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBio1D3 monoclonal antibody reacts with mouse CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells.