missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD19 Rat anti-Mouse, FITC, Clone: eBio1D3 (1D3), eBioscience™

Rat Monoclonal Antibody

$117.48 - $645.48

Specifications

Antigen CD19
Clone eBio1D3 (1D3)
Host Species Rat
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122044 Life Technologies
LS11019385
500μg Each for $381.48
509471 Life Technologies
LS11019381
50μg Each for $117.48
509474 Life Technologies
LS11019386
1mg Each for $645.48
501129692 Life Technologies
LS11019382
100μg Each for $209.88
Specifications

Description

The eBio1D3 monoclonal antibody reacts with mouse CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells.
Description & Specifications

Specifications

CD19
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBio1D3 (1D3)
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL