Please login to your online account to display your discounted pricing

CD19 Rat anti-Mouse, Clone: eBio1D3 (1D3), Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$129.05 - $477.05

Specifications

Antigen CD19
Clone eBio1D3 (1D3)
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014250

 
life technologies
LS16019381
50μg Each for $129.05

5014251

 
life technologies
LS16019385
500μg Each for $477.05
Description & Specifications

Specifications

Antigen CD19
Clone eBio1D3 (1D3)
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The eBio1D3 monoclonal antibody reacts with mouse CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells.