missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD19 Rat anti-Mouse, Biotin, Clone: eBio1D3 (1D3), eBioscience™

Rat Monoclonal Antibody

$104.28 - $619.08

Specifications

Antigen CD19
Clone eBio1D3 (1D3)
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011460 Life Technologies
LS13019381
50μg Each for $104.28
5011461 Life Technologies
LS13019382
100μg Each for $196.68
5011462 Life Technologies
LS13019385
500μg Each for $341.88
5011463 Life Technologies
LS13019386
1mg Each for $619.08
Specifications

Description

The eBio1D3 monoclonal antibody reacts with mouse CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells.
Description & Specifications

Specifications

CD19
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
eBio1D3 (1D3)
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer