missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD19 Mouse anti-Human, PerCP-eFluor 710, Clone: J3-129, eBioscience™

Mouse Monoclonal Antibody

$209.88 - $421.08

Specifications

Antigen CD19
Clone J3-129
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015931 Life Technologies
LS46019741
25 Tests Each for $209.88
5015932 Life Technologies
LS46019742
100 Tests Each for $421.08
Specifications

Description

This J3-129 monoclonal antibody reacts with human CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development, excluding terminally differentiated plasma cells. However, follicular dendritic cells express CD19. CD21, CD81, Leu13, MHC class II, and CD19 together form a multi-molecular complex that associates with the B cell receptor. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and B cell proliferation.
Description & Specifications

Specifications

CD19
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
J3-129
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test