Please login to your online account to display your discounted pricing

CD19 Mouse anti-Human, PE-eFluor 610, Clone: HIB19, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $442.25

Specifications

Antigen CD19
Clone HIB19
Host Species Mouse
Gene Alias Leu-12
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124741

 
life technologies
LS61019941
25 tests Each for $172.55

501124742

 
life technologies
LS61019942
100 tests Each for $442.25
Description & Specifications

Specifications

Antigen CD19
Clone HIB19
Host Species Mouse
Gene Alias Leu-12
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The HIB19 monoclonal antibody reacts with human CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, Leu13, MHC class II, and CD19 form a multimolecular complex that associates with BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. The SJ25C1 antibody and the HIB19 monoclonal antibody recognize overlapping epitopes.