missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD19 Mouse anti-Human, PE-Cy5, Clone: HIB19, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $341.88

Specifications

Antigen CD19
Clone HIB19
Host Species Murine
Gene Alias Leu-12
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014024 Life Technologies
LS15019941
25 Tests Each for $157.08
5014025 Life Technologies
LS15019942
100 Tests Each for $341.88
Specifications

Description

The HIB19 monoclonal antibody reacts with human CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, Leu13, MHC class II, and CD19 form a multimolecular complex that associates with BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. The SJ25C1 antibody and the HIB19 monoclonal antibody recognize overlapping epitopes.
Description & Specifications

Specifications

CD19
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HIB19
Leu-12
Flow Cytometry
PE-Cy5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test