Please login to your online account to display your discounted pricing

CD19 Mouse anti-Human, PE, Clone: SJ25C1, eBioscience™

Mouse Monoclonal Antibody

$130.50 - $323.35

Specifications

Antigen CD19
Clone SJ25C1
Host Species Mouse
Gene Alias Leu-12
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245259

 
life technologies
LS12019841
25 tests Each for $130.50

50245260

 
life technologies
LS12019842
100 tests Each for $323.35
Description & Specifications

Specifications

Antigen CD19
Clone SJ25C1
Host Species Mouse
Gene Alias Leu-12
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The SJ25C1 monoclonal antibody reacts with human CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express this molecule. CD19, along with CD21, CD81, Leu13, and MHC class II, form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux, and proliferation of B cells. The SJ25C1 antibody and the HIB19 monoclonal antibody recognize overlapping epitopes.