Please login to your online account to display your discounted pricing

CD19 Mouse anti-Human, Clone: LC1, eBioscience™

Mouse Monoclonal Antibody

$72.50 - $72.50

Specifications

Antigen CD19
Clone LC1
Host Species Murine
Gene Alias Leu-12
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122634

 
life technologies
LS14019080
25μg Each for $72.50
Description & Specifications

Specifications

Antigen CD19
Clone LC1
Host Species Murine
Gene Alias Leu-12
Species Reactivity Human
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The LC1 monoclonal antibody reacts with human CD19, a 95kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, Leu13, MHC class II, and CD19 form a multi-molecular complex that associates with BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. The antibody recognizes a sequence near the carboxy terminus.