missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD185 (CXCR5) Rat anti-Mouse, PE-Cy7, Clone: SPRCL5, eBioscience™

Rat Monoclonal Antibody

$196.68 - $460.68

Specifications

Antigen CD185 (CXCR5)
Clone SPRCL5
Host Species Rat
Gene Alias C-X-C Chemokine Receptor Type 5, BLR1, MDR15
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123441 Life Technologies
LS25718580
25μg Each for $196.68
501123442 Life Technologies
LS25718582
100μg Each for $460.68
Specifications

Description

This SPRCL5 monoclonal antibody reacts with Mouse CD185. CD185, which is also known as C-X-C chemokine receptor 5 (CXCR5) and Burkitt lymphoma receptor 1 (BLR1), is a seven transmembrane G protein-coupled receptor originally identified in Burkitt’s lymphoma. In peripheral blood, CXCR5 is expressed on B cells, CD4+ T cells (but not Th1 or Th2 cells), as well as on a subpopulation of memory (CD45RO+) T cells. CXCR5+ circulating T cells are in a resting state and migrate to the lymph nodes due to expression of CCR7 and CD62L. In tonsil, CXCR5 is expressed on nearly all CD4+ cells along with CD45RO and such activation markers as CD69 and ICOS. Tonsillar CXCR5+ cells have been shown to induce antibody production when co-cultured with B cells, thus supporting their role in providing B cell help. Furthermore, this chemokine receptor plays a critical role in lymphocyte trafficking, in particular T cell migration into the B cell follicles of germinal centers in response to CXCL13, making CXCR5 an established marker of follicular helper T cells.

This SPRCL5 does not see a formaldehyde fixed epitope, therefore staining should be done prior to fixation.
Description & Specifications

Specifications

CD185 (CXCR5)
Rat
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
SPRCL5
C-X-C Chemokine Receptor Type 5, BLR1, MDR15
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer