Please login to your online account to display your discounted pricing

CD185 (CXCR5) Rat anti-Mouse, PE, Clone: SPRCL5, eBioscience™

Rat Monoclonal Antibody

$187.05 - $477.05

Specifications

Antigen CD185 (CXCR5)
Clone SPRCL5
Host Species Rat
Gene Alias C-X-C Chemokine Receptor Type 5, BLR1, MDR15
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122385

 
affymetrix
12718582
100μg Each for $477.05

501128646

 
affymetrix
12718580
25μg Each for $187.05
Description & Specifications

Specifications

Antigen CD185 (CXCR5)
Clone SPRCL5
Host Species Rat
Gene Alias C-X-C Chemokine Receptor Type 5, BLR1, MDR15
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer

This SPRCL5 monoclonal antibody reacts with Mouse CD185. CD185, which is also known as C-X-C chemokine receptor 5 (CXCR5) and Burkitt lymphoma receptor 1 (BLR1), is a seven transmembrane G protein-coupled receptor originally identified in Burkitt’s lymphoma. In peripheral blood, CXCR5 is expressed on B cells, CD4+ T cells (but not Th1 or Th2 cells), as well as on a subpopulation of memory (CD45RO+) T cells. CXCR5+ circulating T cells are in a resting state and migrate to the lymph nodes due to expression of CCR7 and CD62L. In tonsil, CXCR5 is expressed on nearly all CD4+ cells along with CD45RO and such activation markers as CD69 and ICOS. Tonsillar CXCR5+ cells have been shown to induce antibody production when co-cultured with B cells, thus supporting their role in providing B cell help. Furthermore, this chemokine receptor plays a critical role in lymphocyte trafficking, in particular T cell migration into the B cell follicles of germinal centers in response to CXCL13, making CXCR5 an established marker of follicular helper T cells.

This SPRCL5 does not see a formaldehyde fixed epitope, therefore staining should be done prior to fixation.