Please login to your online account to display your discounted pricing

CD185 (CXCR5) Mouse anti-Human, PE-Cy7, Clone: MU5UBEE, eBioscience™

Mouse Monoclonal Antibody

$191.40 - $487.20

Specifications

Antigen CD185 (CXCR5)
Clone MU5UBEE
Host Species Mouse
Gene Alias C-X-C chemokine receptor type 5, BLR1, MDR15
Species Reactivity Human, Non-human primate, Rhesus Ma
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123449

 
life technologies
LS25918541
25 tests Each for $191.40

501128663

 
life technologies
LS25918542
100 tests Each for $487.20
Description & Specifications

Specifications

Antigen CD185 (CXCR5)
Clone MU5UBEE
Host Species Mouse
Gene Alias C-X-C chemokine receptor type 5, BLR1, MDR15
Species Reactivity Human, Non-human primate, Rhesus Ma
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The MU5UBEE monoclonal antibody reacts with human and non-human primate CD185. CD185, which is also known as C-X-C chemokine receptor 5 (CXCR5) and Burkitt lymphoma receptor 1 (BLR1), is a seven transmembrane G protein-coupled receptor originally identified in Burkitt’s lymphoma. In peripheral blood, CD185 is expressed on B cells, CD4+ T cells (but not Th1 or Th2 cells), as well as on a subpopulation of memory (CD45RO+) T cells. Circulating CD185+ T cells are in a resting state and migrate to the lymph nodes due to expression of CCR7 and CD62L. In tonsil, CD185 is expressed on nearly all CD4+ cells together with CD45RO and activation markers such as CD69 and ICOS. Tonsillar CD185+ cells have been shown to induce antibody production when co-cultured with B cells, thus supporting their role in providing help to B cells. Furthermore, this chemokine receptor plays a critical role in lymphocyte trafficking, in particular T cell migration into the B cell follicles of germinal centers in response to CXCL13, making CD185 an established marker of follicular helper T cells.

The MU5UBEE monoclonal antibody also crossreacts with Rhesus macaque.