Please login to your online account to display your discounted pricing

CD185 (CXCR5) Mouse anti-Human, Clone: MU5UBEE, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen CD185 (CXCR5)
Clone MU5UBEE
Host Species Murine
Species Reactivity Human, Non-human primate, Rhesus Ma
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122776

 
life technologies
LS14918580
25μg Each for $143.55

501122777

 
life technologies
LS14918582
100μg Each for $361.05
Description & Specifications

Specifications

Antigen CD185 (CXCR5)
Clone MU5UBEE
Host Species Murine
Species Reactivity Human, Non-human primate, Rhesus Ma
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MU5UBEE monoclonal antibody reacts with Mouse CD185. CD185, which is also known as C-X-C chemokine receptor 5 (CXCR5) and Burkitt lymphoma receptor 1 (BLR1), is a seven transmembrane G protein-coupled receptor originally identified in Burkitt’s lymphoma. In peripheral blood, CXCR5 is expressed on B cells, CD4+ T cells (but not Th1 or Th2 cells), as well as on a subpopulation of memory (CD45RO+) T cells. CXCR5+ circulating T cells are in a resting state and migrate to the lymph nodes due to expression of CCR7 and CD62L. In tonsil, CXCR5 is expressed on nearly all CD4+ cells along with CD45RO and such activation markers as CD69 and ICOS. Tonsillar CXCR5+ cells have been shown to induce antibody production when co-cultured with B cells, thus supporting their role in providing B cell help. Furthermore, this chemokine receptor plays a critical role in lymphocyte trafficking, in particular T cell migration into the B cell follicles of germinal centers in response to CXCL13, making CXCR5 an established marker of follicular helper T cells.