missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD184 (CXCR4) Rat anti-Mouse, eFluor 450, Clone: 2B11, eBioscience™

Rat Monoclonal Antibody

$174.24 - $402.60

Specifications

Antigen CD184 (CXCR4)
Clone 2B11
Host Species Rat
Gene Alias Fusin, C-X-C chemokine receptor type 4, SDF-1 Receptor
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124239 Life Technologies
LS48999180
25μg Each for $174.24
501124240 Life Technologies
LS48999182
100μg Each for $402.60
Specifications

Description

The 2B11 monoclonal antibody reacts with mouse and human CXCR4, also known as CD184, Fusin, LESTR, or HUMSTR. Fusin is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. In addition it has been shown to play a role in lymphocyte migration. The ligand for CXCR4 is SDF1. CXCR4 is required for maintenance of granulocyte progenitors in the bone marrow and essential for B cell development. Expression of CXCR4 is different between mice and human. In the thymus CXCR4 is restricted to CD4+CD8+ cells while in the spleen predominant expression is found on B lymphocytes.

The monoclonal antibody 2B11 recognizes the amino terminus of CXCR4 and has been reported to have functional activity; it does not block SDF1a but causes internalization of the receptor.
Description & Specifications

Specifications

CD184 (CXCR4)
Rat
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
2B11
Fusin, C-X-C chemokine receptor type 4, SDF-1 Receptor
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer