missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD184 (CXCR4) Rat anti-Mouse, Biotin, Clone: 2B11, eBioscience™

Rat Monoclonal Antibody

$117.48 - $289.08

Specifications

Antigen CD184 (CXCR4)
Clone 2B11
Host Species Rat
Gene Alias Fusin
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012313 Life Technologies
LS13999180
25μg Each for $117.48
5012314 Life Technologies
LS13999182
100μg Each for $289.08
Specifications

Description

The 2B11 monoclonal antibody reacts with mouse and human CXCR4, also known as CD184, Fusin, LESTR, or HUMSTR. Fusin is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. In addition it has been shown to play a role in lymphocyte migration. The ligand for CXCR4 is SDF1. CXCR4 is required for maintenance of granulocyte progenitors in the bone marrow and essential for B cell development. Expression of CXCR4 is different between mice and human. In the thymus CXCR4 is restricted to CD4+CD8+ cells while in the spleen predominant expression is found on B lymphocytes.

The monoclonal antibody 2B11 recognizes the amino terminus of CXCR4 and has been reported to have functional activity; it does not block SDF1a but causes internalization of the receptor.
Description & Specifications

Specifications

CD184 (CXCR4)
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
2B11
Fusin
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer