missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD184 (CXCR4) Mouse anti-Human, PE, Clone: 12G5, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $368.28

Specifications

Antigen CD184 (CXCR4)
Clone 12G5
Host Species Murine
Gene Alias Fusin
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011355 Life Technologies
LS12999941
25 Tests Each for $157.08
5011356 Life Technologies
LS12999942
100 Tests Each for $368.28
Specifications

Description

The 12G5 monoclonal antibody reacts with human CD184 (CXCR4), also termed Fusin, LESTR, or HUMSTR. CD184 is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. CD184 is expressed predominantly on naïve T cell subsets of peripheral blood and is rapidly upregulated by PHA and IL-2 stimulation. The 12G5 antibody shows partial inhibition of chemotaxis and calcium influx induced by SDF-1 (the natural ligand of CD184), blocks CD4-independent HIV-2 infection, and blocks CD4-dependent infection by some T-tropic HIV-1 isolates.
Description & Specifications

Specifications

CD184 (CXCR4)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
12G5
Fusin
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer