missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD184 (CXCR4) Mouse anti-Human, Biotin, Clone: 12G5, eBioscience™

Mouse Monoclonal Antibody

$104.28 - $289.08

Specifications

Antigen CD184 (CXCR4)
Clone 12G5
Host Species Murine
Gene Alias Fusin
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012321 Life Technologies
LS13999980
25μg Each for $104.28
5012322 Life Technologies
LS13999982
100μg Each for $289.08
Specifications

Description

The 12G5 monoclonal antibody reacts with human CD184 (CXCR4), also termed Fusin, LESTR, or HUMSTR. CD184 is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. CD184 is expressed predominantly on naïve T cell subsets of peripheral blood and is rapidly upregulated by PHA and IL-2 stimulation. The 12G5 antibody shows partial inhibition of chemotaxis and calcium influx induced by SDF-1 (the natural ligand of CD184), blocks CD4-independent HIV-2 infection, and blocks CD4-dependent infection by some T-tropic HIV-1 isolates.
Description & Specifications

Specifications

CD184 (CXCR4)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
12G5
Fusin
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL